missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-Naphthalenesulfonate 98.0+%, TCI America™
Supplier: TCI America N0015500G
Specifications
| Sodium 1-Naphthalenesulfonate | |
| White | |
| C10H7NaO3S | |
| 500 g | |
| HIEHAIZHJZLEPQ-UHFFFAOYSA-M | |
| sodium;naphthalene-1-sulfonate | |
| 23661867 | |
| ≥98.0% (HPLC,T) |
| 130-14-3 | |
| 5.98 (5% w/v H2O soln. 20°C) | |
| MFCD00064964 | |
| sodium 1-naphthalenesulfonate, sodium naphthalene-1-sulfonate, 1-naphthalenesulfonic acid sodium salt, unii-mai7v3c3pn, sodium naphthalenesulphonate, sodium naphthalene sulfonate, 1-naphthalenesulfonic acid, sodium salt, alpha salt, sodium alpha-naphthalenesulfonate, mai7v3c3pn | |
| C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)[O-].[Na+] | |
| 230.213 | |
| 230.21 | |
| Crystalline Powder |
Chemical Identifiers
| 130-14-3 | |
| 230.213 | |
| HIEHAIZHJZLEPQ-UHFFFAOYSA-M | |
| 23661867 | |
| C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)[O-].[Na+] |
| C10H7NaO3S | |
| MFCD00064964 | |
| sodium 1-naphthalenesulfonate, sodium naphthalene-1-sulfonate, 1-naphthalenesulfonic acid sodium salt, unii-mai7v3c3pn, sodium naphthalenesulphonate, sodium naphthalene sulfonate, 1-naphthalenesulfonic acid, sodium salt, alpha salt, sodium alpha-naphthalenesulfonate, mai7v3c3pn | |
| sodium;naphthalene-1-sulfonate |
Safety and Handling
EINECSNumber : (4)-0466
TSCA : Yes