missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Naphthalenesulfonate 98.0+%, TCI America™

Catalog No. N001525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N001525G 25 g
N0015100G 100 g
N0015500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N001525G Supplier TCI America Supplier No. N001525G

Chemical Identifiers

CAS 130-14-3
Molecular Formula C10H7NaO3S
Molecular Weight (g/mol) 230.213
MDL Number MFCD00064964
InChI Key HIEHAIZHJZLEPQ-UHFFFAOYSA-M
Synonym sodium 1-naphthalenesulfonate, sodium naphthalene-1-sulfonate, 1-naphthalenesulfonic acid sodium salt, unii-mai7v3c3pn, sodium naphthalenesulphonate, sodium naphthalene sulfonate, 1-naphthalenesulfonic acid, sodium salt, alpha salt, sodium alpha-naphthalenesulfonate, mai7v3c3pn
PubChem CID 23661867
IUPAC Name sodium;naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)[O-].[Na+]

Specifications

Color White
pH 5.98 (5% w/v H2O soln. 20°C)
Quantity 25 g
Formula Weight 230.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Naphthalenesulfonate
EINECSNumber (4)-0466
TSCA Yes