missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Hexanesulfonate 98.0+%, TCI America™

Catalog No. p-7131351
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03445G 5 g
I034425G 25 g
I0344100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I03445G Supplier TCI America Supplier No. I03445G
Only null left

Chemical Identifiers

CAS 2832-45-3
Molecular Formula C6H13NaO3S
Molecular Weight (g/mol) 188.22
MDL Number MFCD00007542
InChI Key QWSZRRAAFHGKCH-UHFFFAOYSA-M
Synonym sodium 1-hexanesulfonate, sodium hexane-1-sulfonate, sodium hexanesulfonate, 1-hexanesulfonic acid sodium salt, 1-hexanesulfonic acid, sodium salt, 1-hexanesulfonic acid, sodium salt 1:1, 1-hexane sulfonic acid sodium salt, hexyl sodium sulfonate, 1-hexanesulfonate, sodium, hexanesulfonic acid na-salt
PubChem CID 23677630
IUPAC Name sodium hexane-1-sulfonate
SMILES [Na+].CCCCCCS([O-])(=O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 188.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 1-Hexanesulfonate [Reagent for Ion-Pair Chromatography]
EINECSNumber (2)-3786
TSCA Yes