missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-Hexanesulfonate Monohydrate, 99%

Catalog No. p-7049966
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3219009 10 g
AA3219018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3219009 Supplier Thermo Scientific Chemicals Supplier No. 03219009
Only null left

CAS: 207300-91-2 | C6H15NaO4S | 206.232 g/mol

Sodium 1-hexanesulfonate monohydrate is an ion pair reagent, which is used for the analysis of small organic compounds, pharmaceutical products and metabolites using high-performance liquid chromatography (HPLC). It is an efficient catalyst for the preparation of alpha-aminophosphonates by the coupling of aldehydes/ketone, an amine and triethyl phosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207300-91-2
Molecular Formula C6H15NaO4S
Molecular Weight (g/mol) 206.232
MDL Number MFCD00149549
InChI Key CLCGFJYKZGFGSQ-UHFFFAOYSA-M
Synonym 1-hexanesulfonic acid sodium salt monohydrate, sodium hexane-1-sulfonate hydrate, sodium 1-hexanesulfonate monohydrate, n-1-hexanesulfonic acid, sodium 1-hexanesulfonate hydrate, sodium n-hexanesulfonate monohydrate, potassium hexane-1-sulfonate hydrate, sodium hexane-1-sulphonate monohydrate, sodium hexane-1-sulfonate-water 1/1/1, 1-hexanesulfonic acid,sodium salt, hydrate 1:1:1
PubChem CID 23696962
IUPAC Name sodium;hexane-1-sulfonate;hydrate
SMILES CCCCCCS(=O)(=O)[O-].O.[Na+]

Specifications

Color White
Linear Formula CH3(CH2)5SO3Na·H2O
Quantity 10 g
Beilstein 3727014
Solubility Information Slightly soluble in water
Formula Weight 206.24 (188.22 Anhydrous)
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Sodium 1-hexanesulfonate monohydrate
EINECSNumber 220-601-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only