missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-dodecanesulfonate, 99%

Catalog No. p-7020765
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1282903 1 g
AAA1282909 10 g
AAA1282914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1282903 Supplier Thermo Scientific Chemicals Supplier No. A1282903
Only null left

CAS: 2386-53-0 | C12H25NaO3S | 272.379 g/mol

Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-dodecanesulfonate is used as a ion-associating reagent for HPLC for analysis of peptides and proteins. It acts as an ion-pair reagent, which increases the retention of thiabendazole on HPLC column during the determination of thiabendazole in fruits like citrus and banana. It is used in the synthesis of graphene-nanocomposites. It is also used as surfactant, flotation agent, foaming agent, penetrating agent and emulsifiers. It is involved in the preparation of 1-Chloro-dodecane.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2386-53-0
Molecular Formula C12H25NaO3S
Molecular Weight (g/mol) 272.379
MDL Number MFCD00007527
InChI Key DAJSVUQLFFJUSX-UHFFFAOYSA-M
Synonym sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
PubChem CID 23665726
IUPAC Name sodium;dodecane-1-sulfonate
SMILES CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 3919536
Solubility Information Soluble in water.
Formula Weight 272.39
Percent Purity 99%
Chemical Name or Material Sodium 1-dodecanesulfonate
Hazard Category H500
EINECSNumber 219-200-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only