missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sodium 1-dodecanesulfonate, 98%

Catalog No. p-7237350
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3659003 1 g
AAH3659009 10 g
AAH3659014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3659003 Supplier Thermo Scientific™ Supplier No. H3659003
Only null left

Chemical Identifiers

CAS 2386-53-0
Molecular Formula C12H25NaO3S
Molecular Weight (g/mol) 272.38
MDL Number MFCD00007527
InChI Key DAJSVUQLFFJUSX-UHFFFAOYSA-M
Synonym 1-Dodecanesulfonic acid sodium salt;
IUPAC Name sodium dodecane-1-sulfonate
SMILES [Na+].CCCCCCCCCCCCS([O-])(=O)=O

Specifications

Melting Point >300°
Physical Form Solid
Quantity 1 g
Linear Formula CH{3}(CH{2}){1}{1}SO{3}Na
Formula Weight 272.39
Percent Purity 98%
ShelfLife 5 Years
EINECSNumber 219-200-6