Learn More
Socorex Qualitix™ Autoclave Pouches from DWK Life Sciences™
Autoclave pouches made with durable material
Supplier: Socorex 3210.1936
Description
- Quick content identification
- Large self-sealing strip, easy to close
- Chevron top seal for easy opening
- Suitable for steam and EO gas sterilization
- Two sterilization indicators
Chemical Identifiers
| ISO 17665-06, ISO 11135 CE | |
| Steam and EO Gas Sterilization | |
| Autoclave Pouch |
Specifications
| ISO 17665-06, ISO 11135 CE | |
| Steam and EO Gas Sterilization | |
| 190 x 360 mm | |
| Autoclave Pouch |