Learn More
Socorex Qualitix™ Autoclave Pouches from DWK Life Sciences™
Autoclave pouches made with durable material
Supplier:  Socorex 3210.0926
 View more versions of this product
Catalog No. 12141531
Only null left
Description
Description
  • Quick content identification
  • Large self-sealing strip, easy to close
  • Chevron top seal for easy opening
  • Suitable for steam and EO gas sterilization
  • Two sterilization indicators
Specifications
Chemical Identifiers
ISO 17665-06, ISO 11135 CE
Steam and EO Gas Sterilization
Autoclave Pouch
Specifications
ISO 17665-06, ISO 11135 CE
Steam and EO Gas Sterilization
90 x 260 mm
Autoclave Pouch
Product Suggestions
Product Suggestions