missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNTG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140012
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5140012 Supplier Invitrogen™ Supplier No. PA5140012
Only null left

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ILRFYTAQDG TDWLRAVSAN IRELTLQNMK MANKCCSPSD QVVHMGWVNE KLQGADSSQT FRPKFLALK Highest antigen sequence indentity to the following orthologs: Rat - 76%, Mouse - 77%.

Syntrophin gamma 2 encodes a protein belonging to the syntrophin family. Syntrophins are cytoplasmic peripheral membrane proteins that bind to components of mechanosenstive sodium channels and the extreme carboxy-terminal domain of dystrophin and dystrophin-related proteins. The PDZ domain of this protein product interacts with a protein component of a mechanosensitive sodium channel that affects channel gating. Absence or reduction of this protein product has been associated with Duchenne muscular dystrophy. There is evidence of alternative splicing yet the full-length nature of these variants has not been described.

Specifications

Antigen SNTG2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNTG2
Gene Accession No. Q9NY99
Gene Alias 2210008K22Rik; 9530013L23; BB121248; G2SYN; Gamma-2-syntrophin; gamma2-syntrophin; MGC133174; SNTG2; SYN5; syntrophin; syntrophin 5; syntrophin gamma 2; syntrophin, gamma 2; syntrophin-5
Gene Symbols SNTG2
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SNTG2 (Product #RP-109083)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54221
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less