missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNRK Polyclonal Antibody

Description
Immunogen sequence: VPQDLEDDLT ATPLSHATVP QSPARAADSV LNGHRSKGLC DSAKKDDLPE LAGPALSTVP PASLKPTASG RKCLFRVEED EEEDEEDKKP MSLSTQVVLR RKPSVTNRLT SRKSAPVLNQ IFEEGESDDE FDMDENLPPK LSRLKM Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.
SNRK may play a role in hematopoietic cell proliferation or differentiation. SNRK is the potential mediator of neuronal apoptosis.
Specifications
Specifications
| Antigen | SNRK |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.10 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SNRK |
| Gene Accession No. | Q63553, Q8VDU5, Q9NRH2 |
| Gene Alias | 2010012F07Rik; AI448042; AW547029; E030034B15; HSNFRK; KIAA0096; mKIAA0096; R74830; RGD:69653}; SNF related kinase; SNF-1 related kinase; SNF1-related kinase; SNF-related serine/threonine-protein kinase; SNF-related serine/threonine-protein kinase; LOW QUALITY PROTEIN: SNF-related serine/threonine-protein kinase; SNFRK; SNRK; snrk {ECO:0000312 |
| Gene Symbols | SNRK |
| Show More |