missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNaPshot™ Multiplex Kit
Description
Use this system to screen and confirm SNPs, detect minor sequence variations, fingerprint bacterial artificial chromosomes.
Order Info
Each
Specifications
Specifications
| Accession Number | Capillary, 61cm Capillary Length, Sequencing, Room Temperature, For 310 Genetic Analyzer |
| Description | SNaPshot™ Multiplex Kit |
| No. of Reactions | 1000 |
For research use only