missing translation for 'onlineSavingsMsg'
Learn More

SNaPshot™ Multiplex Kit

Catalog No. 4323154
Encompass
Change view
Click to view available options
No. of Reactions:
1000
5000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
4323154 1000
4323155 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4323154 Supplier Thermo Fisher Scientific Supplier No. 4323154

Use this system to screen and confirm SNPs, detect minor sequence variations, fingerprint bacterial artificial chromosomes.

Use this system to screen and confirm SNPs, detect minor sequence variations, fingerprint bacterial artificial chromosomes.

Order Info

Each

Specifications

Accession Number Capillary, 61cm Capillary Length, Sequencing, Room Temperature, For 310 Genetic Analyzer
Description SNaPshot™ Multiplex Kit
No. of Reactions 1000

For research use only