missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Snap Ring Polypropylene Autosampler Microvial

Catalog No. 111010037
Change view
Click to view available options
Volume (Metric):
0.1 mL
0.5 mL
0.75 mL
No. per Pack:
100
1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) No. per Pack
111010037 0.75 mL 1000
111011843 0.1 mL 100
111011968 0.5 mL 100
111012028 0.75 mL 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111010037 Supplier MilliporeSigma™ Supelco™ Supplier No. 27254

Autoclavable, easy to maintain. A vial is used to store or as a closure type.

Specifications

Material Natural Polypropylene
Volume (Metric) 0.75 mL
No. per Pack 1000
Style Large opening, (6 mm)