missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube
Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.
Supplier: Eppendorf™ 022363697
Description
- The hinged Eppendorf Safe-Lock lid prevents unintended opening during incubation and storage.
- g-Safe™ technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30,000 x g, preventing sample loss in the event of tube breakage and providing enhanced safety when handling hazardous samples.
- Manufactured without the use of slip agents, plasticizers, and biocides, substances known to leach from plastic consumables into the sample and adversely impact bio-assay results.
- Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures.
- Precise lid sealing minimizes evaporation rates during long-term storage.
- The large frosted lid and side surface facilitate easy labeling.
- Ensured functionality in a temperature range from -86 °C to 100 °C.
- Autoclavable when open (121°C, 20 min.) for effective sterilization.
- Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur™ quality, ensuring tested purity.
Order Info
Assorted packages include 100 each of natural, blue, green, red and yellow
Chemical Identifiers
| Autoclavable | |
| Polypropylene | |
| -86°C to +100°C | |
| 0.02 oz. | |
| Cylindrical |
| 0.5 mL | |
| -122.8°F to +212°F | |
| Yes | |
| Safe-lock Lid | |
| Locking Snap-cap Microtube |
Specifications
| Polypropylene | |
| 0.5 mL | |
| Polypropylene | |
| 500 Tubes ( 5 Bags x 100 Tubes) | |
| -86°C to +100°C | |
| Eppendorf Quality | |
| 0.02 oz. | |
| 1.3 in. | |
| 30,000 x g | |
| 9 mm | |
| 33 mm |
| Autoclavable | |
| Assorted | |
| 500 Pk. | |
| -122.8°F to +212°F | |
| Non-sterile | |
| Yes | |
| Safe-lock Lid | |
| Conical | |
| 0.25 mL | |
| Cylindrical | |
| Locking Snap-cap Microtube |