missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube

Catalog No. 0540223
Change view
Click to view available options
Color:
Amber
Assorted
Blue
Clear
Green
Red
Yellow
Purity or Quality Grade:
Biopur™ (Sterile)
Eppendorf Quality
PCR Clean (Non-sterile)
Packaging:
50 Tubes, Individually Packed
500 Tubes ( 5 Bags x 100 Tubes)
500 Tubes (1 Bag x 500 Tubes)
Max. RCF:
22,000 x g
30,000 x g
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Purity or Quality Grade Packaging Max. RCF
0540223 Yellow Eppendorf Quality 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
0540293 Clear PCR Clean (Non-sterile) 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
0540219 Amber PCR Clean (Non-sterile) 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
0540218 Clear Eppendorf Quality 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
0540220 Blue Eppendorf Quality 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
0540221 Green Eppendorf Quality 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
0540222 Red Eppendorf Quality 500 Tubes (1 Bag x 500 Tubes) 30,000 x g
054024 Assorted Eppendorf Quality 500 Tubes ( 5 Bags x 100 Tubes) 30,000 x g
0540224A Clear Biopur™ (Sterile) 50 Tubes, Individually Packed 22,000 x g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 0540223 Supplier Eppendorf™ Supplier No. 022363671

Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.

  • The hinged Eppendorf Safe-Lock lid prevents unintended opening during incubation and storage.
  • g-Safe™ technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30,000 x g, preventing sample loss in the event of tube breakage and providing enhanced safety when handling hazardous samples.
  • Manufactured without the use of slip agents, plasticizers, and biocides, substances known to leach from plastic consumables into the sample and adversely impact bio-assay results.
  • Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures.
  • Precise lid sealing minimizes evaporation rates during long-term storage.
  • The large frosted lid and side surface facilitate easy labeling.
  • Ensured functionality in a temperature range from -86 °C to 100 °C.
  • Autoclavable when open (121°C, 20 min.) for effective sterilization.
  • Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur™ quality, ensuring tested purity.

Order Info

Assorted packages include 100 each of natural, blue, green, red and yellow

Specifications

Closure Material Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 0.5 mL
Color Yellow
Material Polypropylene
No. per Pack 500 Pk.
Packaging 500 Tubes (1 Bag x 500 Tubes)
Temperature Range (English) -122.8°F to +212°F
Temperature Range (Metric) -86°C to +100°C
Sterility Non-sterile
Purity or Quality Grade Eppendorf Quality
Graduated Yes
Capacity (English) 0.02 oz.
Closure Type Safe-lock Lid
Length (English) 1.3 in.
Bottom Shape Conical
Max. RCF 30,000 x g
Graduations 0.25 mL
Diameter (Metric) 9 mm
Shape Cylindrical
Length (Metric) 33 mm
Product Type Locking Snap-cap Microtube
Show More Show Less