missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Safe-Lock Tubes 0.5 mL - Microtube
Description
- The hinged Eppendorf Safe-Lock lid prevents unintended opening during incubation and storage.
- g-Safe™ technology, dependent on the Safe-Lock Tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30,000 x g, preventing sample loss in the event of tube breakage and providing enhanced safety when handling hazardous samples.
- Manufactured without the use of slip agents, plasticizers, and biocides, substances known to leach from plastic consumables into the sample and adversely impact bio-assay results.
- Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures.
- Precise lid sealing minimizes evaporation rates during long-term storage.
- The large frosted lid and side surface facilitate easy labeling.
- Ensured functionality in a temperature range from -86 °C to 100 °C.
- Autoclavable when open (121°C, 20 min.) for effective sterilization.
- Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur™ quality, ensuring tested purity.
Order Info
Assorted packages include 100 each of natural, blue, green, red and yellow
Specifications
Specifications
| Closure Material | Polypropylene |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 0.5 mL |
| Color | Amber |
| Material | Polypropylene |
| No. per Pack | 500 Pk. |
| Packaging | 500 Tubes (1 Bag x 500 Tubes) |
| Temperature Range (English) | -122.8°F to +212°F |
| Temperature Range (Metric) | -86°C to +100°C |
| Sterility | Non-sterile |
| Show More |