missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ 1.5 mL Flex-Tube™ - Microtube
Description
- Easy-to-use ergonomic design featuring a user-friendly tube lid.
- Durable polypropylene construction capable of withstanding chemicals, mechanical stress, and extreme temperatures.
- g-Safe™ technology ensures stability up to 30,000 x g, preventing sample loss during centrifugation and enhancing safety with hazardous samples.
- Free from slip agents, plasticizers, and biocides, eliminating substances that may compromise bio-assay results.
- Clear graduation marks allow for visual volume verification during pipetting.
- Convenient frosted lid and side surface facilitate easy labeling.
- Functional across a wide temperature range (-86 °C to 100 °C) for versatile applications.
- Autoclavable up to 121 °C for 20 minutes, ensuring effective sterilization.
- Available in both Eppendorf Quality and PCR Clean purity grades with lot certification.
Specifications
Specifications
| Cap Style | Snap Cap |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 1.5 mL |
| Color | Red |
| Material | Polypropylene |
| For Use With (Application) | For centrifuging or storing samples |
| Includes | Flex lid |
| No. per Pack | 500 Pk. |
| Packaging | 1 Bag x 500 Tubes |
| Temperature Range (English) | -122.8°F to +212°F |
| Show More |