missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ 1.5 mL Flex-Tube™ - Microtube

Catalog No. 0540233
Change view
Click to view available options
Color:
Amber
Blue
Clear
Green
Red
Yellow
Purity or Quality Grade:
Eppendorf Quality
PCR Clean
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Purity or Quality Grade
0540233 Green Eppendorf Quality
05402 Clear Eppendorf Quality
0540231 Amber Eppendorf Quality
0540235 Yellow Eppendorf Quality
0540234 Red Eppendorf Quality
0540232 Blue Eppendorf Quality
0540296 Clear PCR Clean
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 0540233 Supplier Eppendorf™ Supplier No. 022363557
Only null left

Flex-Tube™ (1.5 mL) are designed for seamless one-handed operation with an easy-to-open, securely closing tube design. Ideal for sample preparation, centrifugation, and storage, these tubes will effortlessly integrate into your workflows for efficient and reliable performance.

  • Easy-to-use ergonomic design featuring a user-friendly tube lid.
  • Durable polypropylene construction capable of withstanding chemicals, mechanical stress, and extreme temperatures.
  • g-Safe™ technology ensures stability up to 30,000 x g, preventing sample loss during centrifugation and enhancing safety with hazardous samples.
  • Free from slip agents, plasticizers, and biocides, eliminating substances that may compromise bio-assay results.
  • Clear graduation marks allow for visual volume verification during pipetting.
  • Convenient frosted lid and side surface facilitate easy labeling.
  • Functional across a wide temperature range (-86 °C to 100 °C) for versatile applications.
  • Autoclavable up to 121 °C for 20 minutes, ensuring effective sterilization.
  • Available in both Eppendorf Quality and PCR Clean purity grades with lot certification.

Specifications

Cap Style Snap Cap
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Green
Material Polypropylene
For Use With (Application) For centrifuging or storing samples
Includes Flex lid
No. per Pack 500 Pk.
Packaging 1 Bag x 500 Tubes
Temperature Range (English) -122.8°F to +212°F
Temperature Range (Metric) -86°C to +100°C
Sterility Non-sterile
Purity or Quality Grade Eppendorf Quality
Graduated Yes
Capacity (English) 0.05 oz.
Closure Type Snap Cap
Length (English) 1.5 in.
Bottom Shape Conical
Max. RCF 30,000 x g
Graduations 0.5 mL
Diameter (Metric) 10 mm
Quantity 500 Tubes
Shape Cylindrical
Length (Metric) 38 mm
Product Type Microtube
Show More Show Less