missing translation for 'onlineSavingsMsg'
Learn More

Motic SMZ171-TLED Stereo and Camera Bundle

Catalog No. 01257889
Change view
Click to view available options
Model:
SMZ-171-TLED w/ BMH4000X Camera Bundle
SMZ-171-TLED with BTI10 Camera Bundle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
01257889 SMZ-171-TLED w/ BMH4000X Camera Bundle
01257488 SMZ-171-TLED with BTI10 Camera Bundle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01257889 Supplier Motic Supplier No. 1100501300053

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The SMZ-171 is an advanced stereo microscope for biomedical work and industrial quality control.

  • Greenough Zoom Optical System
  • 45° stereomicroscope head
  • 6.7:1 Zoom Ratio
  • Diopter Adjustment: ±5

Specifications

Includes BMH4000X Camera Bundle
Voltage 100/240 V
Magnification Power 0.75X to 5X
Objective Types Zoom Stereo
Weight (Metric) 6.2 kg
Dimensions (L x W x H) 303 x 239 x 405 mm
Product Type Microscope with Digital Camera
Model SMZ-171-TLED w/ BMH4000X Camera Bundle
Head Style Trinocular
Camera Type Moticam BMH4000X
Lighting Type LED, Transmitted Illumination
Eyepiece N-WF 10X/23 mm, with diopter adjustment on both
Working Distance 110 mm
Show More Show Less