missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMS Recombinant Rabbit Monoclonal Antibody (23GB4640)

Catalog No. MA553781
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553781 20 μL
MA553780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553781 Supplier Invitrogen™ Supplier No. MA553781
Only null left

Rabbit Recombinant Monoclonal Antibody

SMS Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Catalyzes the production of spermine from spermidine and decarboxylated S-adenosylmethionine (dcSAM).

Specifications

Antigen SMS
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4640
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SMS
Gene Accession No. P52788
Gene Alias AI427066; Gy; gyro; MRSR; SMS; Spermidine aminopropyltransferase; spermine synthase; SpmST; SPMSY; SpS; SRS
Gene Symbols SMS
Host Species Rabbit
Immunogen A synthesized peptide derived from human Spermine synthase (100-150AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6611
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less