missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMS Monoclonal Antibody (K52015_5E8), Invitrogen™
Mouse Monoclonal Antibody
$1,601.06 - $2,821.92
Specifications
| Antigen | SMS |
|---|---|
| Clone | K52015_5E8 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: MAAARHSTLD FMLGAKADGE TILKGLQSIF QEQGMAESVH TWQDHGYLAT YTNKNGSFAN LRIYPHGLVL LDLQSYDGDA QGKEEIDSIL NKVEERMKEL SQDSTGRVKR LPPIVRGGAI DRYWPTADGR LVEYDIDEVV YDEDSPYQNI KILHSKQFGN ILILSGDVNL AESDLAYTRA IMGSGKEDYT GKDVLILGGG DGGILCEIVK LKPKMVTMVE IDQMVIDGCK KYMRKTCGDV LDNLKGDCYQ VLIEDCIPVL KRYAKEGREF DYVINDLTAV PISTSPEEDS TWEFLRLILD LSMKVLKQDG KYFTQGNCVN LTEALSLYEE QLGRLYCPVE FSKEIVCVPS YLELWVFYTV WKKAKP
SMS Monoclonal antibody specifically detects SMS in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| SMS | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AI427066; Gy; gyro; MRSR; SMS; Spermidine aminopropyltransferase; spermine synthase; SpmST; SPMSY; SpS; SRS | |
| Recombinant human SMS (immunogen range is 1-366). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| SMS |
| K52015_5E8 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P52788 | |
| 6611 | |
| IgG1 | |
| Protein A | |
| Antibody |