missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Smooth Muscle Differentiation Supplement (SMDS)

Catalog No. S0085
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0085 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0085 Supplier Gibco™ Supplier No. S0085

Smooth Muscle Differentiation Supplement (SMDS)

A sterile, concentrated (100X) solution intended for use with Medium 231 to enhance differentiation of human smooth muscle cells. Each 5 ml bottle of SMDS contains the correct amount of differentiation supplement for a 500 ml bottle of Medium 231. When a 500 ml bottle of medium is supplemented with SMDS, the final concentrations of the components in the supplemented medium are:
  • fetal bovine serum, 1% v/v
  • heparin, 30μg/ml
  • Specifications

    Content And Storage minus 20 degrees C
    Cell Type Smooth Muscle Cells
    Concentration 100 X
    Form Liquid
    Product Type Differentiation Supplement
    Sterility Sterile
    For Use With (Application) Cell Differentiation
    Product Line Cascade Biologics
    Quantity 5 mL
    Shipping Condition Dry Ice
    Species Human
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.