missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-nitro-1,3-propanediol, 98.0-100.5%, Spectrum™ Chemical

Catalog No. 18608199
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608199 25 kg Amber Glass Bottle
18608194 25 g Amber Glass Bottle
18608195 100 g Amber Glass Bottle
18608196 1 kg Amber Glass Bottle
18608197 5 kg Amber Glass Bottle
18608198 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608199 Supplier Spectrum Chemical Mfg Cor Supplier No. B229425KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52-51-7
Molecular Formula C3H6BrNO4
Molecular Weight (g/mol) 199.99
InChI Key LVDKZNITIUWNER-UHFFFAOYSA-N
IUPAC Name 2-bromo-2-nitropropane-1,3-diol
SMILES OCC(Br)(CO)[N+]([O-])=O

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 kg
Percent Purity 98.0 to 100.5%
Grade Reagent
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.