missing translation for 'onlineSavingsMsg'
Learn More

SMARCD1/BAF60a Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501560785
Encompass
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501560785 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501560785 Supplier Bethyl Laboratories Inc Supplier No. A301595AT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine

The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and has sequence similarity to the yeast Swp73 protein. Two transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen SMARCD1/BAF60a
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SMARCD1
Gene Accession No. Q61466, Q96GM5
Gene Alias BAF60A, CRACD1, Rsc6p, 60 kDa BRG-1/Brm-associated factor subunit A, BRG1-associated factor 60A, SWI/SNF complex 60 kDa subunit A, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily D member 1, Swp73-like protein, chromatin remodeling complex BAF60A s
Gene Symbols SMARCD1
Host Species Rabbit
Immunogen Between 250 and 300
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6602, 83797
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less