missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SMAD6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01676639
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01676639 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01676639 Supplier Invitrogen™ Supplier No. PA141026
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

This antibody is predicted to react with equine based on sequence homology. Suggested positive control: HeLa or NIH-3T3 whole cell lysate. Cross reacts with New World Monkey.

SMAD6 (mothers against decapentaplegic homolog 6) is an antagonist of signaling by TGF-beta (transforming growth factor) type 1 receptor superfamily members; has been shown to inhibit selectively BMP (bone morphogenetic proteins) signaling by competing with the co-SMAD SMAD4 for receptor-activated SMAD1. SMAD6 is an inhibitory SMAD (I-SMAD) or antagonistic SMAD.

Specifications

Antigen SMAD6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SMAD6
Gene Accession No. O35182, O43541
Gene Alias AOVD2; b2b390Clo; HsT17432; MAD homolog 6; mad homolog 7; Madh6; Madh7; Mothers against decapentaplegic homolog 6; mothers against decapentaplegic, drosophila, homolog of, 6; mothers against DPP homolog 6; Msmad6; SMAD; SMAD 6; SMAD family member 6; SMAD SMAD; SMAD, mothers against DPP homolog 6; SMAD6
Gene Symbols SMAD6
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human SMAD6 (between amino acids 30-150).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101103068, 17130, 367100, 4091
Target Species Human, Mouse, Monkey, Sheep, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less