missing translation for 'onlineSavingsMsg'
Learn More

Calcium Chloride, Anhydrous, Flakes / Granules, Technical, Spectrum™ Chemical

Catalog No. 18609466
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609466 100 lb. Poly Bottle
18609463 500 g Poly Bottle
18609464 2.5 kg Poly Bottle
18609465 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18609466 Supplier Spectrum Chemical Mfg Cor Supplier No. C1097100LBBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10043-52-4

Specifications

Physical Form Flakes or Powder
CAS Max % 1
Quantity 100 lb.
Packaging Poly Bottle
Molecular Weight (g/mol) 110.98
Percent Purity 100%
Grade Technical
Chemical Name or Material Calcium Chloride, Anhydrous, Flakes / Granules

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.