Learn More
Motic™ SM7 CMO Stereo Zoom Binocular Microscope
The SM7 is a modular CMO stereo microscope for use in biomedicine and industrial advanced research work. This microscope currently provides a 112x magnification. It can be used on a boom stand as well for imaging large parts on a table.
Supplier:  Motic™ 1100201800061
 View more versions of this product
Catalog No. 01674219
Only null left
Description
Description
The SM7 features an optical system with high resolution, true color reproduction, and low distortion.
  • Advanced ergonomic design features have been carefully implemented to maximize efficient usage and minimize operator fatigue
  • Ideal positioning and angles of each module, compact size, and high-quality provide a user-friendly environment for any task
  • Variety of objective lenses and eyepieces allows the user to tailor the SM7 to their specific purpose or task
  • Customized stand and digital application guarantee the best resolution no matter the environment
  • Galilean Optical Zoom System, denoting an Infinity Optical System, allows for high flexibility
  • Optical setup can be varied by easily exchanging eyepieces and/or objectives (for best possible mage results, Plan-Apochromatic objective 1X (WD 81 mm) is available)
  • Standard eyepieces 10X/22 offer diopter adjustment range of ±5, allowing to insert reticles for visual measurements
  • 8x to 56x zoom range provided makes execution of any task much easier
  • More comfortable, longer working distance
  • Live color reproduction and high-resolution power minimize distortion and aberration, offering a clear image on a wide range of samples
  • Observation tube is adaptable to any situation: trino head is suitable for video port digital imaging applications (60° inclination tube is also available to meet special use and custom needs)
  • Careful polyurethane material selection in zoom knobs and focus dials assures comfort during any operation
  • To avoid repetitive zooming changes, a click stop function has been implemented to improve work efficiency
  • High Eye Point eyepiece design provides comfortable viewing when operating with glasses or low visual acuity
Specifications
Chemical Identifiers
Galilean
367 x 290 x 372 mm
Binocular
WF 10X/22 mm
Specifications
Microscope with Plain Stand
Plan Achromatic 0.8X to 5.6X (10-step)
Binocular
90 mm
Galilean
367 x 290 x 372 mm
WF 10X/22 mm
Product Suggestions
Product Suggestions