missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLITRK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520477
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520477 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520477 Supplier Invitrogen™ Supplier No. PA520477
Only null left

Rabbit Polyclonal Antibody

A suggested positive control is NIH-3T3 cell lysate. PA5-20477 can be used with blocking peptide PEP-0597.

SLIT and NTRK-like family 5 (Slitrk5) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the protein family. Like every other Slitrk protein except Slitrk1, overexpression of Slitrk5 inhibited neurite outgrowth in cultured neurons, suggesting that these proteins are involved in the control of neurite outgrowth.

Specifications

Antigen SLITRK5
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLITRK5
Gene Accession No. O94991, Q810B7
Gene Alias 2610019D03Rik; bA364G4.2; KIAA0918; leucine rich repeat containing 11; leucine-rich repeat-containing protein 11; LRRC11; SLIT and NTRK like family member 5; SLIT and NTRK-like family, member 5; SLIT and NTRK-like protein 5; slit and trk like gene 5; Slitrk5
Gene Symbols SLITRK5
Host Species Rabbit
Immunogen A 14 amino acid peptide from near the center of human Slitrk5.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26050, 306152, 75409
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less