missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ Slimline Flat White Light Panel
GREENER_CHOICE

Catalog No. 01257297
Change view
Click to view available options
Wavelength:
352 nm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength
01257297 352 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01257297 Supplier Syngene™ Supplier No. WLP2014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Slimline Flat White Light Panel is ideal for visualising visible proteins, coomassie blue, etc.

  • For use with G:Box Chemi XX6 and XX9 systems
  • Size: 20 x 14 cm
  • With power supply for darkroom or benchtop use.

Specifications

Intensity Fixed
Quantity 1 Ea.
Wavelength 352 nm
For Use With (Equipment) Syngene Gbox XX6 and XX9