missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ SlideMate™ Adhesion Microscope Slides

SlideMate™ Adhesion Microscope Slides offers crisp, clean printing. Designed for use on SlideMate thermal printers. Reduces identification issues common to handwritten labels when paired with SlideMate printers. Comes 144 slides per gross; 10 gross per case.

Supplier:  Epredia™ TT6041IPS82181

 View more versions of this product

Catalog No. 22042951


Only null left
Explore available promotions

Description

Description

  • Manufactured in the USA; SlideMate slides are smooth, three-quarter inch white, blue, green, pink or yellow tab slides validated to work with the SlideMate AS slide printer.
  • The tab is impervious to virtually all common laboratory chemicals and reagents.
  • Available in adhesion and non-adhesion formulas. SlideMate Plus Adhesion Slides feature adhesion coating for routine clinical applications.
  • Epredia SlideMate slides are made from extra-white glass, manufactured to high quality standards. The clarity achieved during the manufacturing process helps ensure high levels of transparency to aid in accurate diagnoses.
  • SlideMate slides come pre-cleaned, flat with finely ground edges. Each package comes wrapped to help protect against humidity and moisture. The packaging is also designed to assist with loading the slides into SlideMate printer hoppers to prevent contamination due to handling.
Specifications

Specifications

Glass
144 Pk.
1440/Cs.
75 mm
Green
90°
1 in.
1 mm
Ground
SlideMate printers
25 mm
Adhesion
3 in.
75 x 25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions