Learn More
United Scientific™ Slide Storage Rack, For 100 Slides
Catalog No. S16053
Change view
Click to view available options
:
Slide Capacity: 100
Catalog No.
S16053 Slide Capacity: 100
1 options
Catalog No. S16053 Supplier United Scientific™ Supplier No. 62501
Only null left
Sturdy and convenient way to store slides.
  • Plastic (ABS) slide storage boxes feature a removable slide tray with 100 numbered slots that hold one or two slides each.
  • Clear hinged lid allows easy view of labels
  • Slides can be stored vertically for easy identification or bar code reading
  • Not Autoclavable
  • 3 x 9 x 3.5 in.
Specifications
Type Slide Storage Box
Slide Capacity 100
For Use With (Equipment) Microscope Slides