Learn More
United Scientific™ Slide Storage Box, Plastic
Catalog No. S16016
Change view
Click to view available options
Slide Capacity:
100
25
50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Slide Capacity
S16016 50
S15961 25
S15856 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S16016 Supplier United Scientific™ Supplier No. SBP050
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Sturdy and convenient way to store slides.
  • Sturdy polystyrene slide boxes designed to hold standard 3 x 1 in. microscope slides.
  • The bottom section has numbers engraved for easy reference, and the inside of the lid carries a numbered slide index.
  • Available in 25, 50, or 100 slide capacity.
Specifications
Type Slide Storage Box
Slide Capacity 50
For Use With (Equipment) Microscope Slides
Warning: Cancer and reproductive harm