Learn More
United Scientific™ Slide Mailer With Cap, PP, Neutral
Convenient way to transfer and store slides while keeping them separated.
Supplier:  United Scientific™ 62606
 View more versions of this product
Catalog No. S012574
Only null left
Explore available promotions
Description
Description
  • A convenient and safe solution for holding and transferring microscope slides. The inside walls feature slotted channels to keep slides separated and to make removal easier. The lids close securely to keep the slides safe.
  • Accommodates microscope slides in two sizes:
    • 75.0 mm x 25.0 mm x (0.8 to 1.2 mm)
    • 76.0 mm x 26.0 mm x (0.8 to 1.2 mm)
  • Securely hold slides: 3-place option with screw cap, and 5-place option with hinge cap
  • Durable Polypropylene
  • Holds two common slide sizes
Specifications
Specifications
Slide Mailer
5
Polypropylene
Product Suggestions
Product Suggestions