missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 3.5K MWCO

Catalog No. p-7226489
Change view
Click to view available options
Sample Volume (Metric):
0.5 mL
3 mL
12 mL
30 mL
Quantity:
1 Kit (10 Cassettes)
1 Kit (8 Cassettes)
10 Cassettes
6 Cassettes
8 Cassettes
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume (Metric) Quantity
PI66333 0.5 mL 10 Cassettes
PI66335 0.5 mL 1 Kit (10 Cassettes)
PI66330 3 mL 10 Cassettes
PI66332 3 mL 1 Kit (10 Cassettes)
PI66110 12 mL 8 Cassettes
PI66107 12 mL 1 Kit (8 Cassettes)
PI66130 30 mL 6 Cassettes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. PI66333 Supplier Thermo Scientific™ Supplier No. 66333
Only null left

Remove salts and contaminants, and maximize sample recovery using this semi-permeable membrane with a 3,500-Dalton protein MW cut-off (MWCO) cassette that is easier to use than traditional dialysis tubing.

Slide-A-Lyzer Dialysis Cassettes (3.5K MWCO) facilitate simple and effective removal of salts and small contaminants from proteins and other macromolecules larger than 3,500 Daltons. They can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration.

Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample loss during the dialysis procedure. Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
Fast dialysis—flat cassette chamber with two membranes provides high surface area-to-volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
High recovery—rectangular cassette design maximizes recovery of entire sample volume from any one of the four corner injection ports
Four cassette sizes—select the cassette that best suits your sample volume (0.5 to 30 mL)
Color-coded—pink cassette frame specifies cassettes with the 3,500 MW pore size membrane

Membrane specifications:
Composition: regenerated cellulose synthesized by the viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: none
Sulfur content: 0.1–0.15%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL
• 30 mL = 12 to 30 mL

Specifications

Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 3.5 kDa
Product Line Slide-A-Lyzer
Quantity 10 Cassettes
Sample Volume (Metric) 0.5 mL