missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Accessory Float Buoys and Syringes

Catalog No. pi66432
Change view
Click to view available options
Capacity (Metric):
For 0.5 to 3 mL Cassettes
For 12 mL Cassettes
1 mL
5 mL
20 mL
Product Type:
Cassette Float Buoy
Cassette Syringe
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Product Type
PI66432 For 12 mL Cassettes Cassette Float Buoy
PI66430 For 0.5 to 3 mL Cassettes Cassette Float Buoy
PI66490 5 mL Cassette Syringe
PI66493 20 mL Cassette Syringe
PI66494 1 mL Cassette Syringe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PI66432 Supplier Thermo Scientific™ Supplier No. 66432
Only null left

Slide-A-Lyzer Float Buoys and Syringes are available to support a variety of dialysis cassettes for volumes of 0.1 to 20 mL.

Slide-A-Lyzer Dialysis Cassette Syringes are designed for efficient loading of samples into Slide-A-Lyzer cassettes and removal of sample after dialysis. Cassette Float Buoy accessories are for use with Slide-A-Lyzer Dialysis Cassettes used to remove low molecular weight contaminants and perform buffer exchange, desalting, and concentration.

Minimize sample loss into and out of Slide-A-Lyzer Dialysis Cassettes using Slide-A-Lyzer Dialysis Cassette Syringes for 1-mL to 20-mL sample volumes. Slide-A-Lyzer Cassette Float Buoys are designed for floating 0.5- to 12-mL cassettes in dialysate.

Features of Slide-A-Lyzer Cassette Accessories include:
• Syringes are appropriately sized for different sample volumes
• Buoys keep stir bar from tearing dialysis membrane
• Matched to Slide-A-Lyzer Cassette for a secure fit and minimization of sample loss
• Chemically inert

Specifications

Sufficient For Floating one 12 mL Cassette with each buoy
Content And Storage Store at room temperature.
Membrane Dialysis
Format Cassette, Accessory
Capacity (Metric) For 12 mL Cassettes
Product Line Slide-A-Lyzer
Quantity 8 Buoys
Sample Volume (Metric) 12 mL
Product Type Cassette Float Buoy