missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Devices, 10K MWCO

Catalog No. PI69570
Change view
Click to view available options
Product Type:
Dialysis Unit
Dialysis Unit, 15 mL Conical Tube
Dialysis Unit, 50 mL Conical Tube
Dialysis Unit, Float Buoy
Dialysis Unit, Microcentrifuge Tubes
Quantity:
1 Kit (10 Devices)
25 Devices
250 Devices
50 Devices
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
PI69570 Dialysis Unit 50 Devices
PI69572 Dialysis Unit 250 Devices
PI69574 Dialysis Unit, Microcentrifuge Tubes 1 Kit (10 Devices)
PI69576 Dialysis Unit, Float Buoy 1 Kit (10 Devices)
PI88401 Dialysis Unit, 15 mL Conical Tube 25 Devices
PI88404 Dialysis Unit, 50 mL Conical Tube 25 Devices
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. PI69570 Supplier Thermo Scientific™ Supplier No. 69570

Perform sample buffer exchange with these 10,000-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.

Slide-A-Lyzer MINI Dialysis Devices, 10K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules greater than 10 kDa in volumes from 10 μL to 2 mL.

Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices. Slide-A-Lyzer MINI Dialysis Devices are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.

Features of Slide-A-Lyzer MINI Dialysis Devices, 10K MWCO:
Three convenient sizes—dialysis devices for 0.1, 0.5, and 2 mL samples fit standard 1.5-mL micro-centrifuge, 15- and 50-mL conical tubes, respectively
Excellent sample recoveries—low-binding plastic and small membrane surface area minimize sample loss compared to filtration and resin systems
One-step protocol—pipette sample into the MINI device and place in tube containing the dialysis buffer; no laborious assembly, device preparation, or expensive equipment are required
100% leak-tested—innovative design does not permit 'wicking' that can occur in home-made devices
Time savings—performs desalting of nucleic acids and proteins (>95% recovery) in less than 15 minutes, which improves sample resolution, quantification, and conjugation

These 10K MWCO dialysis devices are available in three sizes (0.1, 0.5, and 2 mL capacities), and are designed to allow easy sample addition and removal using a standard laboratory pipette. The MINI devices are effective and convenient for accomplishing typical sample desalting or buffer exchange in 4–8 hours. Unlike other small sample separation devices, there is no equipment to assemble, no need for syringe adapters, and no laborious steps necessary to manipulate the small volume sample. The regenerated cellulose membrane is compatible with a number of common chemicals and buffers.

The 0.1 mL MINI dialysis devices are designed to be capped (caps included) and inserted into standard 1.5 mL micro-centrifuge tubes. Alternatively, as many as 25 MINI devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple samples inside a large beaker. The 0.5- and 2-mL MINI dialysis devices fit in and are capped by inserting them into the included 15- and 50-mL conical tubes, respectively. The tubes can be securely placed in standard laboratory racks or shakers; this feature saves space, minimizes the risk of spills or contamination, and eliminates the need for floats or large beakers of buffer. Using these MINI dialysis devices, low molecular weight contaminant removal and buffer exchange and desalting can be accomplished within 4–8 hours with efficiencies, rates, and recoveries very similar to conventional dialysis using a large dialysate volume.

Using the 0.1 mL MINI device, 100 μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes. As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume; temperature and agitation rates; the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes; and the pH, ionic strength, and solvent characteristic of the buffers being used.

Specifications

Product Type Dialysis Unit
Product Line Slide-A-Lyzer
MWCO 10 kDa
Membrane Cellulose Membrane, Dialysis
Format MINI Dialysis Device
Purification Target Buffer Exchange, Protein
Sample Volume (Metric) 0.1 mL
Kit Contents Dialysis Units
Quantity 50 Devices
Content And Storage Store at room temperature
With what chemicals is the Slide-A-Lyzer MINI Dialysis Unit compatible?

The Slide-A-Lyzer MINI Dialysis Unit is compatible with alcohols, ketones, esters, oxides, and solvents containing nitrogen.
Alkalis: 1N sodium hydroxide or 1N ammonium hydroxide may cause pore swelling or shrinkage; not recommended to be used with 1N potassium hydroxide
Acids: Will survive weak acids (100% acetic or formic acids), or ~25% phosphoric or sulfuric acids; it should not be used with >25% strong acids, although a weak solution of a strong acid may work

What percent of my sample should I expect to lose during dialysis when using the Slide-A-Lyzer MINI Dialysis Unit?

In general, polypropylene and regenerated cellulose have very low nonspecific binding properties. The percent recovery of a sample will depend on the unique properties of the sample and on the initial concentration of the sample.

What materials are used to manufacture the Slide-A-Lyzer MINI Dialysis Unit?

The membrane is regenerated cellulose and the plastic is a polypropylene copolymer. Both materials have very low protein and nucleic acid binding properties.

How long does it take to dialyze with the Slide-A-Lyzer MINI Dialysis Unit?

The Slide-A-Lyzer MINI Dialysis Unit can convert 100 µL of pH 2.8 buffer to pH 9.4 dialyzing against 1 liter bicarbonate buffer, pH 9.4 in less than 10 minutes.

How do I use the Slide-A-Lyzer MINI Dialysis Unit?

- Wearing gloves, apply sample (0.01-0.1 mL) using a pipette.
- OPTIONAL: Soak the Slide-A-Lyzer MINI Dialysis Unit to remove trace contaminants.
- Cap the Slide-A-Lyzer MINI Dialysis Unit (to minimize loss) and snap it into the float (sold separately).
- Insert the float into the beaker containing the dialysate.
- Recover sample.

How do I remove trace contaminants from the Slide-A-Lyzer MINI Dialysis Unit?

To remove glycerol, dialyze for 15 minutes against 1 liter of deionized water. Glycerol content is less than 3% in the 3K unit, approximately 15% in the 7K unit and 23% in the 10K unit. To remove metals, dialyze for 15 minutes against 1 liter of 1 mM EDTA. Metals present in the 3K, 7K and 10K unit include 2 ppb iron, 5 ppb magnesium, 1.5 ppb nickel, 0.2 ppb zinc, 0.2 ppb copper, 0.5 ppb chromium and 0.3 ppb cadmium. To remove sulfur, dialyze for 15 minutes against 1 liter of deionized water. Sulfur content should be minimal, less than 0.1%. Thermo Scientific quality control checks for sulfur on the membrane prior to the unit's assembly.

How do I obtain optimum performance with the Slide-A-Lyzer MINI Dialysis Unit?

Always wear gloves when touching a Slide-A-Lyzer MINI Dialysis Unit If desired, soak the Slide-A-Lyzer MINI Dialysis Unit to remove contaminants For best recovery, tilt the tube and collect the sample in the bottom corner of the device.

How can I use the Slide-A-Lyzer MINI Dialysis Unit in my laboratory?

The Slide-A-Lyzer MINI Dialysis Unit is ideal for:

- Salt reduction Buffer exchange (<10 min for pH change 2.8 to 9.5)
- Removal of small molecular weight compounds
- Micro-conjugation (reagent addition or sample evaluation anytime)
- Equilibrium dialysis (microtube, paraffin to adjust depth)
- Concentration (in a microtube concentrates @ 35-45 µl/hr; use at least a 3:1 v/v ratio of concentrator to sample.)
- Sample preparation for instrumentation analysis (HPLC, CE, MS, GC, electrophoresis, etc.)
- Sample preparation for proteins, peptides, oligonucleotides, DNA, RNA (~330 MW/base ~660 MW/bp)
- The “scavenger” technique in which a resin, membrane, polymer or protein is placed in a microtube with buffer in order to select a compound to remove from a sample in a Slide-A-Lyzer MINI Dialysis Unit.

If the Slide-A-Lyzer Dialysis Cassette is not reusable, why are there four portholes?

It is possible to add reagents to the Slide-A-Lyzer Dialysis Cassette at different times using different portholes, allow a reaction to occur, and then dialyze in the same Slide-A-Lyzer Dialysis Cassette, reducing sample loss through reduced sample handling.

I lost a lot of my proteins during the dialysis. How can I prevent this?

The dialysis membranes are made of regenerated cellulose, which may cause some molecules to stick nonspecifically resulting in sample loss. The percent of total protein lost is partially dependent on the protein concentration. Protein loss caused by nonspecific binding to the membrane is negligible for concentrated samples (greater than 0.5 mg/mL) but may be significant with dilute protein samples (<0.1 mg/mL). Adding a “carrier” protein such as BSA to dilute protein sample before dialysis will prevent this loss.

After dialysis, my sample volume increased significantly. What can I do to avoid this?

Water moves quickly and easily across the dialysis membrane. When dialyzing a high solute concentration against a dilute dialysis buffer, there will be a net movement of water (and possibly salts) into the dialysis unit through the membrane. Glycerol and some sugars are especially hygroscopic, and as rapidly as they diffuse across the membrane to reach equilibrium, they also significantly affect the osmosis of water across the membrane and so may cause a change in volume of the sample. Take care when dialyzing with large differences in glycerol or sugar concentration between sample and dialysis membrane. Prevent this movement of water and consequent change in sample volume by dialyzing in a “stepwise” fashion, minimizing the difference in water concentration between sample and dialysis buffer at each stage in the dialysis process (see more in Dialysis: An Overview - https://tools.thermofisher.com/content/sfs/brochures/TR0020-Dialysis-overview.pdf).

The smallest sizes of the Slide-A-Lyzer Mini Dialysis Units (10-100 µL) are inserted into a float. If the top of the sample is below the top of the buffer, then hydrostatic pressure of the buffer will force buffer into the unit, increasing the volume and diluting the sample. Making sure that the dialysis membrane is in contact with the buffer and the top of the sample is at or above the level of the buffer will prevent this

Can I re-use my MINI Slide-A-Lyzer device?

We do not recommend re-use.

Can the MINI Slide-A-Lyzer device be used with nucleic acids?

Yes, but we‘d suggest pre-treatment of the Slide-A-Lyzer device with EDTA or other DNase/RNase pre-treatment.

Can I concentrate my solution with the MINI Slide-A-Lyzer device?

Yes, the MINI Slide-A-Lyzer device can be used as a concentrating device as described in the following article: (https://assets.thermofisher.com/TFS-Assets/LSG/brochures/TR0078-SAL-Conc-Solution.pdf). The average concentration rate of 35 µL/hour was observed in house when the sample was in PBS. We recommend using at least a 3:1 v/v ratio of concentrator to sample. In general, increased salt concentration in the sample will reduce the rate of sample concentration.

Are the MINI Slide-A-Lyzer Dialysis Units sterile?

The MINI Slide-A-Lyzer devices are not certified sterile but they are manufactured in a clean environment. HEPA filters keep out contaminants. From molding to packaging, the MINI components have never been touched with ungloved (powder-free latex or nitrile) hands.

How long should I dialyze my sample when using the MINI Slide-A-Lyzer device?

In general, colder solutions take longer to dialyze. Dialysis completion will depend on the composition and volume of sample and dialysate. Simple pH exchange proceeds very rapidly; less than 10 minutes for 100 µL to change from pH 2.8 to pH 9.4. Dialysis of a 1 M salt solution against water proceeds very rapidly (less than 10 minutes for 10 µL). Dialysis of 100 µL of small compounds, 500-1,500 daltons, against a saline solution will be approximately 50% complete in 2-4 hours or 99% or higher complete after overnight when dialyzing against approximately 1 liter of buffer. Dialysis will proceed faster with more frequent buffer changes.

Does the MINI Slide-a-Lyzer device fit within a 50 mL conical tube? Or within a 15 mL conical tube?

Yes, the 0.1 mL devices can be placed into a foam float during dialysis, while the 0.5 mL and 2 mL sizes are capped by inserting them into the included 15 mL and 50 mL conical tubes, respectively.

What materials are the Slide-A-Lyzer MINI Dialysis Units made of?

The MINI Dialysis Units are made of regenerated cellulose membrane, and the cup is made of polypropylene. The Units require 2 minutes‘ hydration time before use.

How much sample will I lose in the Slide-A-Lyzer Dialysis Cassette?

The Slide-A-Lyzer Dialysis Cassette is the same regenerated cellulose as other dialysis tubing so you can expect about the same amount of protein loss as with tubing. As a guide, a 1 mg/mL solution will have a recovery rate of greater than 95%; at 100 mg/mL the recovery drops to 75-80%; and in solutions as dilute 10 µg/mL users may observe only a 50% (or less) recovery.

What conditions do you recommend for dialysis?

Dialyze for the amount of time sufficient to remove low molecular weight compounds for the specific downstream application. A typical dialysis procedure is as follows:

1) Dialyze for 2 hrs at room temperature or 4 degrees C.
2) Change the dialysis buffer and dialyze for another 2 hrs.
3) Change the dialysis buffer and dialyze overnight at 4 degrees C.

Use the dialysis buffer at 200-500 times the volume of the sample. For example, if the starting sample volume is 1 mL, then use a dialysis buffer amount that is equal to 200 mL-500 mL for each dialysis buffer change.

What is the buoy for?

The buoy plays two roles: it suspends the Slide-A-Lyzer Dialysis Cassette above the stirring bar during dialysis; it provides a stand to set the Slide-A-Lyzer Dialysis Cassette in on the bench-top

Is it necessary for me to buy needles and syringes if I already have them?

No, any 18 gauge needle at least one inch long, and syringes of sufficient volume will work.

What is the highest temperature and the pH range that is compatible with the Slide-A-Lyzer Dialysis Cassette?

The Slide-A-Lyzer Dialysis Cassette can be held at 56 degrees C for 2 hours. At temperatures and times above these, the ABS plastic gets soft and can deform. We recommend maintaining the pH range of 5-9.

Do I have to extensively wash or boil the Slide-A-Lyzer Dialysis Cassette before use to remove contaminants?

No. The Slide-A-Lyzer Dialysis Cassette membrane is of a grade used for human kidney dialysis and requires no pretreatment. However, when using small volumes we do recommend pre-wetting the units in deionized water for 30 seconds. Trace metals and 12% glycerol are present in all Slide-A-Lyzer Dialysis Cassettes. During the dialysis process these will dialyze away to virtually no concentration remaining in the cassette.

What materials are used to make Slide-A-Lyzer Dialysis Cassettes and are they compatible with organic solvents?

The plastic casing is made of ABS plastic and the gasket is made from a non-leaching silicone-type material. The membrane is made from regenerated cellulose. We have successfully used 10% solutions of DMF, DMSO, acetonitrile, methanol, hexane, heptane, 100% acetic acid, and 70% ethanol with the device. The units are compatible with 75% acetonitrile and 1% TFA in the sample, however we recommend not dialyzing into a buffer of acetonitrile concentration greater than 10%.

What sizes of the Slide-A-Lyzer Dialysis Cassette are available and in what MWCO?

There are 3.5 K, 7K, 10K, and 20K MWCOs available in 0.1-0.5 mL, 0.5-3 mL, 3-12 mL, and 12-30 mL. These units, especially at the smaller volumes, provide for greater sample recovery and shorter dialysis times than traditional tubing due to their greater surface area-to-volume ratio. For samples smaller than 100 µL, use the Slide-A-Lyzer MINI Dialysis Units and for larger volumes, refer to our SnakeSkin Dialysis Tubing.

Can I reuse the Slide-A-Lyzer Dialysis Cassette?

No, the Slide-A-Lyzer Dialysis Cassette is designed as a single-use, disposable device. Once hydrated, the membrane immediately begins to dry after the sample is removed: and the whole membrane will be dry within a few minutes, changing the molecular weight cut-off (MWCO).

Why doesn‘t the Slide-A-Lyzer Dialysis Cassette leak?

There is an unseen internal gasket which reseals as the needle is withdrawn. The most important items to remember are:

Fill with air first to check that system is airtight
Puncture each porthole only once
Do not allow the membrane to come in contact with the sharp bevel of the needle

What are the advantages of using a cassette for dialysis over traditional tubing methods?

Cassettes have easy handling and secure sample delivery, while traditional tubing is slippery when wet and can be difficult to handle, increasing risk of sample loss during sample addition/removal. Cassettes provide sample protection with a welded membrane and leak-proof cap, helping to minimize the risk of sample loss, whereas with traditional tubing, samples can leak out if the clamp is loose/falls off. Lastly, cassettes are fast and efficient with high surface/volume ratio allowing for more rapid dialysis than conventional tubing.

How is dialysis performed using the Slide-A-Lyzer Cassette (first generation)?

Please see the protocol summary below:

Insert syringe needle through the gasket via one of the corner ports. Inject the sample, withdraw the excess air and remove the syringe.
Attach a float buoy and dialyze. (Buoys also serve as convenient bench-top stands for the cassettes).
Insert empty syringe needle at a second corner port. Inject air to expand the cassette chamber, then withdraw the dialyzed sample.

What is the difference between a pack and a kit for the Slide-A-Lyzer cassettes?

The kit includes float buoys and syringes, and are only available for first-generation products. Please note that floats and syringes can also be purchased separately.

Are these Slide-A-Lyzer cassettes sterile?

We do not test for or claim sterility.

Do you offer gamma-irradiated array Slide-A-Lyzer cassettes?

Yes, we offer gamma-irradiated array Slide-A-Lyzer cassettes for both first-generation products (Cat. No. 66454, 66455, 66453, 66456) and second-generation products (Cat. No. 88250, 882551, 882552, 882553, 882554).

What is the difference between the original and second-generation Slide-A-Lyzer Cassettes?

The first generation cassettes can require a syringe to insert the sample, and may require a float buoy. They can hold 0.5-30 mL solution. The second-generation cassettes can be loaded with a syringe or pipet, are self-floating, and can hold 0.5-70 mL.

What is the difference between your dialysis devices (plates, device, cassettes, flasks)?

Please view our selection table to choose the right dialysis device for your experiment - https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-purification-isolation/protein-dialysis-desalting-concentration/dialysis-products.html.

What is protein dialysis?

Dialysis is the separation of small and large molecules in a solution by selective diffusion through a semi-permeable membrane. It is generally used for larger sample volumes, and can take hours to overnight for complete dialysis.