missing translation for 'onlineSavingsMsg'
Learn More

SLC7A11 Polyclonal Antibody

SLC7A11 Polyclonal Antibody

Supplier:  Thermo Fisher Scientific PA533050

Catalog No. PIPA533050


Only null left
Explore available promotions

Description

Description

SLC7A11 Polyclonal Antibody for IHC (P)

Specifications

Specifications

SLC7A11
Polyclonal
Unconjugated
SLC7A11
9930009M05Rik; AI451155; Amino acid transport system xc-; calcium channel blocker resistance protein CCBR1; CCBR1; cysteine/glutamate transporter; cystine/glutamate transporter; SLC7A11; sodium independent anionic amino acid transport system; solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11; solute carrier family 7 (cationic amino acid transporter, y+ system), member 11; solute carrier family 7 member 11; solute carrier family 7, (cationic amino acid transporter, y+ system) member 11; sut; xCT
Rabbit
Antigen affinity chromatography
RUO
23657
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q9UPY5
SLC7A11
Synthetic 19 amino acid peptide from N-terminus of human SLC7A11.
50 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions