missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Acetate, Powder, Purified, 99%, Spectrum™ Chemical
Specifications
Specifications
| Color | White |
| CAS Max % | 1 |
| Physical Form | Powder, Crystals and/or Chunks |
| Quantity | 100 g |
| Linear Formula | CH3COOAg |
| Solubility Information | Solubility in Water: 10.2 g/L (20°C). |
| InChI Key | WJCVKPGZQKODGL-UHFFFAOYSA-L |
| Molecular Weight (g/mol) | 166.92 |
| Formula Weight | 166.91 g/mol |
| Percent Purity | 99% |
| Show More |
Safety and Handling
| Recommended Storage | Light Sensitive, Store in a dark place |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.