accessibility menu, dialog, popup

UserName

AgCH3OO

CAS RN 563-63-3

IUPAC Name: silver(1+) acetate
Synonyms: silver acetate
Molecular Weight (g/mol): 166.91
Molecular Formula: C2H3AgO2
InChi Key: CQLFBEKRDQMJLZ-UHFFFAOYSA-M
SMILES: [Ag+].CC([O-])=O
Quantity 
  • (3)
  • (3)
  • (1)
Grade 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)