missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC6A18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537221
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537221 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537221 Supplier Invitrogen™ Supplier No. PA537221
Only null left

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 71 kDa. Purity is >95% by SDS-PAGE.

The solute carrier (SLC) family, also known as the neurotransmitter transporter family, is one of the largest transporter families in the human genome. The SLC6 subgroup, which includes the dopamine transporter (DAT), the serotonin transporter (SERT) and the norepinephrine transporter (NET), contains sodium/chloride dependent, high-affinity plasma transport proteins. Sodium/chloride-dependent transporter XTRP2, also known as solute carrier family 6 member 18 (SLC6A18), is a 628 amino acid member of the SLC family. XTRP2 is composed of 12 exons, 11 introns and 8 tandem repeats. Highly expressed in the kidney, SLC6A18 is classified as an orphan transporter.

Specifications

Antigen SLC6A18
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.2
Gene SLC6A18
Gene Accession No. O88576, Q62687, Q96N87
Gene Alias b(0)AT3; B0AT3; D630001K16Rik; FLJ31236; Inactive sodium-dependent neutral amino acid transporter B(0)AT3; renal osmotic stress-induced Na-Cl organic solute cotransporter; Rosit; Slc6a18; sodium and chloride dependent transporter Xtrp2; sodium- and chloride-dependent transporter XTRP2; sodium channel-like protein; Sodium-dependent neutral amino acid transporter B(0)AT3; solute carrier family 6 (neurotransmitter transporter), member 18; solute carrier family 6 (neutral amino acid transporter), member 18; solute carrier family 6 member 18; solute carrier family 6, member 18; system B 0 neutral amino acid transporter AT3; system B(0) neutral amino acid transporter AT3; X transporter protein 2; Xt2; Xtrp2
Gene Symbols SLC6A18
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in human SLC6A18.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22598, 29323, 348932
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less