missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC51A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA526837
Description
SLC51A Polyclonal Antibody for Western Blot, Flow
Essential component of the Ost-alpha/Ost-beta complex, a heterodimer that acts as the intestinal basolateral transporter responsible for bile acid export from enterocytes into portal blood. Efficiently transports the major species of bile acids (taurocholate). Taurine conjugates are transported more efficiently across the basolateral membrane than glycine-conjugated bile acids (By similarity). Can also transport steroids such as estrone 3-sulfate and dehydroepiandrosterone 3- sulfate, therefore playing a role in the enterohepatic circulation of sterols. Able to transport eicosanoids such as prostaglandin E2 (By similarity).Specifications
| SLC51A | |
| Polyclonal | |
| Unconjugated | |
| SLC51A | |
| organic solute transporter alpha; organic solute transporter subunit alpha; organic solute transporter, alpha subunit; OSTA; OSTalpha; OST-alpha; SLC51A; solute carrier family 51 alpha subunit; solute carrier family 51 subunit alpha; solute carrier family 51, alpha subunit | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 200931 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q86UW1 | |
| SLC51A | |
| KLH conjugated synthetic peptide between 298-327 amino acids from the C-terminal region of human OST alpha. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |