missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC4A9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103608
Description
Antibody detects endogenous levels of total SLC4A9.
SLC4A9 Probable apical anion exchanger of the kidney cortex. Belongs to the anion exchanger (TC 2.A.31) family. 3 isoforms of the human protein are produced by alternative splicing. Protein type: Transporter, SLC family; Membrane protein, integral; Transporter; Membrane protein, multi-pass. Cellular Component: apical part of cell; plasma membrane; integral to membrane. Molecular Function: inorganic anion exchanger activity. Biological Process: bicarbonate transport; ion transport; transmembrane transport.Chemical Identifiers
| SLC4A9 | |
| -20°C | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Monkey | |
| SLC4A9 | |
| AE 4; AE4; Anion exchange protein 4; anion exchanger 4; D630003B07; D630024F24Rik; SBC5; SLC4A9; Sodium bicarbonate cotransporter 5; solute carrier family 4 member 9; solute carrier family 4, sodium bicarbonate cotransporter, member 9 | |
| SLC4A9 | |
| Primary | |
| Affinity chromatography |
| 1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96Q91 | |
| 240215, 83697 | |
| A synthesized peptide derived from human SLC4A9(Accession Q96Q91), corresponding to amino acid residues A314-A364. | |
| Antibody |
Specifications
| SLC4A9 | |
| Polyclonal | |
| Unconjugated | |
| SLC4A9 | |
| AE 4; AE4; Anion exchange protein 4; anion exchanger 4; D630003B07; D630024F24Rik; SBC5; SLC4A9; Sodium bicarbonate cotransporter 5; solute carrier family 4 member 9; solute carrier family 4, sodium bicarbonate cotransporter, member 9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 240215, 83697 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96Q91 | |
| SLC4A9 | |
| A synthesized peptide derived from human SLC4A9(Accession Q96Q91), corresponding to amino acid residues A314-A364. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |