missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC4A8 Polyclonal Antibody

Description
SLC4A8 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Mediates electroneutral sodium- and carbonate-dependent choride-HCO3(-) exchange with a Na(+):HCO3(-) stoichiometry of 2:1. Plays a major role in pH regulation in neurons. May be involved in cell pH regulation by transporting HCO3(-) from blood to cell. Enhanced expression in severe acid stress could be important for cell survival by mediating the influx of HCO3(-) into the cells. Also mediates lithium-dependent HCO3(-) cotransport. May be regulated by osmolarity.
Specifications
Specifications
| Antigen | SLC4A8 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | Slc4a8 |
| Gene Accession No. | Q2Y0W8 |
| Gene Alias | AW493845; C230026C11; Electroneutral Na(+)-driven Cl-HCO3 exchanger; electroneutral Na+-driven Cl-HCO3 exchanger; Electroneutral sodium bicarbonate exchanger 1; KIAA0739; k-NBC3; kNBC-3; Na-driven Cl-HCO3 exchanger NDCBE1-A; Na-driven Cl-HCO3 exchanger NDCBE1-B; Na-driven Cl-HCO3 exchanger NDCBE1-C; NBC; NBC3; NDCBE; NDCBE1; RGD:735164}; Slc4a8; slc4a8 {ECO:0000312; sodium bicarbonate cotransporter isoform 3 kNBC-3; solute carrier family 4 (anion exchanger), member 8; solute carrier family 4 member 8; solute carrier family 4, sodium bicarbonate cotransporter, member 8 |
| Gene Symbols | Slc4a8 |
| Show More |