missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC4A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557433
Description
Immunogen sequence: KHSDPHLLER NGEGLSASRH SLRTGLSASN LSLRGESPLS LLLGHLLPSS RAGTPAGSRC TTPVPTPQNS PPSSPSISRL Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 98%.
SLC4A7 regulates intracellular pH and may also play a role in bicarbonate salvage in secretory epithelia. Additionally, SLC4A7 may also have associated sodium channel activity.Chemical Identifiers
| SLC4A7 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| SLC4A7 | |
| bicarbonate transporter; BT; E430014N10Rik; Electroneutral Na/HCO(3) cotransporter; electroneutral sodium bicarbonate cotransporter 1; Na-bicarbonate cotransporter; Nbc; NBC2; NBC2B; NBC3; NBC-like protein; Nbcn1; NBCn1-G; SBC2; SLC4A6; Slc4a7; Sodium bicarbonate cotransporter 2; sodium bicarbonate cotransporter 2b; Sodium bicarbonate cotransporter 3; sodium bicarbonate cotransporter NBCn1-A; sodium bicarbonate cotransporter NBCn1-B; sodium bicarbonate cotransporter NBCn1-c; sodium bicarbonate cotransporter NBCn1-D; sodium bicarbonate cotransporter NBCn1-H; solute carrier family 4 member 7; solute carrier family 4, sodium bicarbonate cotransporter, member 7 | |
| SLC4A7 | |
| Primary | |
| Antigen affinity chromatography |
| 0.1 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6M7 | |
| 9497 | |
| Recombinant protein corresponding to Human SLC4A7. Recombinant protein control fragment (Product #RP-96255). | |
| Antibody |
Specifications
| SLC4A7 | |
| Polyclonal | |
| Unconjugated | |
| SLC4A7 | |
| bicarbonate transporter; BT; E430014N10Rik; Electroneutral Na/HCO(3) cotransporter; electroneutral sodium bicarbonate cotransporter 1; Na-bicarbonate cotransporter; Nbc; NBC2; NBC2B; NBC3; NBC-like protein; Nbcn1; NBCn1-G; SBC2; SLC4A6; Slc4a7; Sodium bicarbonate cotransporter 2; sodium bicarbonate cotransporter 2b; Sodium bicarbonate cotransporter 3; sodium bicarbonate cotransporter NBCn1-A; sodium bicarbonate cotransporter NBCn1-B; sodium bicarbonate cotransporter NBCn1-c; sodium bicarbonate cotransporter NBCn1-D; sodium bicarbonate cotransporter NBCn1-H; solute carrier family 4 member 7; solute carrier family 4, sodium bicarbonate cotransporter, member 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9497 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y6M7 | |
| SLC4A7 | |
| Recombinant protein corresponding to Human SLC4A7. Recombinant protein control fragment (Product #RP-96255). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |