missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC4A11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553730
Description
Immunogen sequence: TNTENEATSG GCVLLHTSRK YLKLKNFKEE IRAHRDLDGF LAQASIVLNE TATSLDNVLR TMLRRFARDP DNNEPNCNLD LLMAMLFTDA GAPMRGKVHL L Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
This gene encodes a voltage-regulated, electrogenic sodium-coupled borate cotransporter that is essential for borate homeostasis, cell growth and cell proliferation. Mutations in this gene have been associated with a number of endothelial corneal dystrophies including recessive corneal endothelial dystrophy 2, corneal dystrophy and perceptive deafness, and Fuchs endothelial corneal dystrophy. Multiple transcript variants encoding different isoforms have been described.Chemical Identifiers
| SLC4A11 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| SLC4A11 | |
| AI503023; bicarbonate transporter related protein 1; bicarbonate transporter-related protein 1; BTR1; CDPD; CDPD1; CHED; CHED2; dJ794I6.2; MGC126418; MGC126419; NABC1; OTTHUMP00000030097; Slc4a11; Sodium bicarbonate transporter-like protein 11; Sodium borate cotransporter 1; sodium-coupled borate cotransporter 1; solute carrier family 4 member 11; solute carrier family 4, sodium bicarbonate transporter-like, member 11; solute carrier family 4, sodium borate transporter, member 11 | |
| SLC4A11 | |
| Primary | |
| Antigen affinity chromatography |
| 0.1 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NBS3 | |
| 83959 | |
| Recombinant protein corresponding to Human SLC4A11. Recombinant protein control fragment (Product #RP-91073). | |
| Antibody |
Specifications
| SLC4A11 | |
| Polyclonal | |
| Unconjugated | |
| SLC4A11 | |
| AI503023; bicarbonate transporter related protein 1; bicarbonate transporter-related protein 1; BTR1; CDPD; CDPD1; CHED; CHED2; dJ794I6.2; MGC126418; MGC126419; NABC1; OTTHUMP00000030097; Slc4a11; Sodium bicarbonate transporter-like protein 11; Sodium borate cotransporter 1; sodium-coupled borate cotransporter 1; solute carrier family 4 member 11; solute carrier family 4, sodium bicarbonate transporter-like, member 11; solute carrier family 4, sodium borate transporter, member 11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83959 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NBS3 | |
| SLC4A11 | |
| Recombinant protein corresponding to Human SLC4A11. Recombinant protein control fragment (Product #RP-91073). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |