missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, Granular, NF, 99.7-100.5%, Spectrum™ Chemical

Catalog No. 18607222
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18607222 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18607222 Supplier Spectrum Chemical Mfg Cor Supplier No. TA105500GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

TA105, 87-69-4, C4H6O6

Spectrum™ Chemical Tartaric Acid, Granular, NF is used as an antioxidant, acidifier, flavor enhancer, stabilizer and sequestering agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

CAS Max % 1
Quantity 500 g
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Poly Bottle
Percent Purity 99.7 to 100.5%
Grade NF