missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC30A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA5150313
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA5150313 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA5150313 Supplier Invitrogen™ Supplier No. PA5150313
Only null left

Rabbit Polyclonal Antibody

SLC30A5 Polyclonal Antibody for IHC, Western Blot

Specifications

Antigen SLC30A5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SLC30A5
Gene Accession No. Q8R4H9, Q8TAD4
Gene Alias 1810010K08Rik; AF233321; hZTL1; SLC30A5; solute carrier family 30 (zinc transporter), member 5; solute carrier family 30 member 5; UNQ863/PRO1879; Zinc transporter 5; zinc transporter like 1; zinc transporter ZTL1; Znt5; ZnT-5; ZNTL1; ZnT-like transporter 1; ZTL1
Gene Symbols SLC30A5
Host Species Rabbit
Immunogen Recombinant Solute Carrier Family 30, Member 5 (SLC30A5) (Asp639-Leu751).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294698, 64924, 69048
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Show More Show Less