missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC27A5 Monoclonal Antibody (4B11C10)

Description
MA5-17175 targets SLC27A5 in indirect ELISA, IHC and WB applications and shows reactivity with Human and Mouse samples. The MA5-17175 immunogen is purified recombinant fragment of human SLC27A5 (amino acids: 508-570 ) expressed in E. Coli. MA5-17175 detects SLC27A5 which has a predicted molecular weight of approximately 75.4kDa.
The protein encoded by this gene is an isozyme of very long-chain acyl-CoA synthetase (VLCS). It is capable of activating very long-chain fatty-acids containing 24- and 26-carbons. It is expressed in liver and associated with endoplasmic reticulum but not with peroxisomes. Its primary role is in fatty acid elongation or complex lipid synthesis rather than in degradation. This gene has a mouse ortholog.
Specifications
Specifications
| Antigen | SLC27A5 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 4B11C10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | SLC27A5 |
| Gene Accession No. | Q4LDG0, Q9Y2P5 |
| Gene Alias | ACSB; ACSVL6; BA-CoA ligase; BACS; BAL; bile acid CoA ligase; bile acid-CoA ligase; Bile acyl-CoA synthetase; cholate--CoA ligase; FACVL3; FATP5; FATP-5; Fatty acid transport protein 5; fatty-acid-Coenzyme A ligase, very long-chain 3; FLJ22987; rBAL-1; Slc27a5; solute carrier family 27 (fatty acid transporter), member 5; solute carrier family 27 member 5; ve; Very long-chain acyl-CoA synthetase homolog 2; very long-chain acyl-CoA synthetase-related protein; very long-chain acyl-Coenzyme A dehydrogenase synthase related; Vlacsr; VLACS-related; VLCSH2; VLCS-H2 |
| Show More |