missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149991
Description
SLC25A37 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
SLC25A37 belongs to the SLC25 family of mitochondrial carrier proteins.Chemical Identifiers
| SLC25A37 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q66H23, Q920G8, Q9NYZ2 | |
| 100153622, 306000, 51312, 67712 | |
| Recombinant Mitoferrin (MFRN) (Pro65-Tyr338). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Pig | |
| SLC25A37 | |
| 1700020E22Rik; 4930513O14Rik; 4930526G11Rik; AI848481; C330015G08Rik; frascati; HT015; Mfrn; Mfrn1; mitochondrial carrier domain containing protein RGD1359361; mitochondrial iron transporter 1; Mitochondrial solute carrier protein; mitochondrial solute carrier-like protein; mitoferrin; mitoferrin 1; mitoferrin-1; MSC; MSCP; predicted protein of HQ2217; PRO1278; PRO1584; PRO2217; RGD1359361; Slc25a37; solute carrier family 25 (mitochondrial iron transporter), member 37; solute carrier family 25 member 37; solute carrier family 25, member 37 | |
| SLC25A37 | |
| Primary | |
| Protein A |
Specifications
| SLC25A37 | |
| Polyclonal | |
| Unconjugated | |
| SLC25A37 | |
| 1700020E22Rik; 4930513O14Rik; 4930526G11Rik; AI848481; C330015G08Rik; frascati; HT015; Mfrn; Mfrn1; mitochondrial carrier domain containing protein RGD1359361; mitochondrial iron transporter 1; Mitochondrial solute carrier protein; mitochondrial solute carrier-like protein; mitoferrin; mitoferrin 1; mitoferrin-1; MSC; MSCP; predicted protein of HQ2217; PRO1278; PRO1584; PRO2217; RGD1359361; Slc25a37; solute carrier family 25 (mitochondrial iron transporter), member 37; solute carrier family 25 member 37; solute carrier family 25, member 37 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100153622, 306000, 51312, 67712 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q66H23, Q920G8, Q9NYZ2 | |
| SLC25A37 | |
| Recombinant Mitoferrin (MFRN) (Pro65-Tyr338). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat, Pig | |
| Antibody |