missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SLC24A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104164
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104164 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104164 Supplier Invitrogen™ Supplier No. PA5104164
Only null left

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC24A2.

NCKX2, also designated solute carrier family 24, member 2 (SLC24A2), belongs to a family of potassium-dependent sodium/calcium exchangers, all of which contain two large hydrophilic loops and two sets of multiple transmembrane-spanning segments. NCKX2 is expressed in neurons throughout the brain, with high expression found in cone photoreceptors in the retina as well as hippocampal pyramidal cells and the deep nuclei of the cerebellum. A critical component in the process of light adaptation, NCKX2 transports one calcium ion and one potassium ion in exchange for four sodium ions, thereby contolling calcium concentration in the eye during light and darkness. Additionally, NCKX2 regulates calcium levels in other cellular locations that experience rapid calcium fluxes, such as neuronal synapses. Inactivation of NCKX2 is believed to be an important regulatory mechanism in the control of sodium and calcium levels under different physiological conditions.

Specifications

Antigen SLC24A2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Slc24a2
Gene Accession No. O54701, Q9UI40
Gene Alias 2810021B17Rik; 6330417K15Rik; AI847460; Na(+)/K(+)/Ca(2+)-exchange protein 2; Nckx2; potassium-dependent sodium-calcium exchanger; retinal cone Na-Ca+K exchanger; Slc24a2; Sodium/potassium/calcium exchanger 2; solute carrier family 24 (sodium/potassium/calcium exchanger), member 2; solute carrier family 24 member 2; solute carrier family 24, member 2
Gene Symbols Slc24a2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC24A2(Accession Q9UI40), corresponding to amino acid residues P311-L361.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25769, 76376, 84550
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less